Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serine racemase Antibody (OTI2E3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274083MFV500
Description
Serine racemase Monoclonal antibody specifically detects Serine racemase in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Serine racemase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2E3 | |
Flow Cytometry | |
D-serine ammonia-lyase, D-serine dehydratase, EC 4.3.1.17, EC 4.3.1.18, EC 5.1.1.18, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, serine racemase | |
Full length huamn recombinant protein of human SRR (NP_068766) produced in HEK293T cell. | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
63826 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction