Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serine racemase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564916 Supplier Invitrogen™ Supplier No. PA564916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVPLVDALVVP VGGGGMLAGI AITVKALKPS VKVYAAEPSN ADDCYQSKLK GKLMPNLYPP ETIADGVKSS IGLNTWPIIR DLVDDIFTVT EDEIKCATQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 92%.

SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serine racemase
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Srr
Gene Accession No. Q9GZT4
Gene Alias D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs
Gene Symbols Srr
Host Species Rabbit
Immunogen Recombinant Human SRR. Recombinant protein control fragment (Product #RP-105710).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.