Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Serine racemase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597207 Supplier Invitrogen™ Supplier No. PA597207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MCAQYCISFA DVEKAHINIR DSIHLTPVLT SSILNQLTGR NLFFKCELFQ KTGSFKIRGA LNAVRSLVPD ALERKPKAVV THSSGNHGQA LTYAAKLEGI PAYIVVPQTA PDCKKLAIQA YGASIVYCEP SDESRENVAK RVTEETEGIM VHPNQEPAVI AGQGTIALEV LNQVPLVDAL VVPVGGGGML AGIAITVKAL KPSVKVYAAE PSNADDCYQS KLKGKLMPNL YPPETIADGV KSSIGLNTWP IIRDLVDDIF TVTEDEIKCA TQLVWERMKL LIEPTAGVGV AAVLSQHFQT VSPEVKNICI VLSGGNVDLT SSITWVKQAE RPASYQSVSV; Positive Samples: Rat brain.

SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serine racemase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Srr
Gene Accession No. Q76EQ0, Q9GZT4, Q9QZX7
Gene Alias D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs
Gene Symbols Srr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human SRR (NP_0687661).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27364, 303306, 63826
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.