Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Monoclonal Antibody (5HT-H209), Invitrogen™

Catalog No. PIMA512111
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512111 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512111 Supplier Thermo Scientific Supplier No. MA512111

Mouse Monoclonal Antibody

MA5-12111 targets Serotonin in IHC (P) applications and shows reactivity with Human samples. The MA5-12111 immunogen is 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin).

Serotonin (also known as 5-hydroxytryptamne or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotoinin is widely distributed in the central and peripheral nervous system, and also in some non-nueral tissues such as platelets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5HT-H209
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene Alias 5-HT; 5-hydroxytryptamne
Host Species Mouse
Immunogen 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin)
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.