Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Serotonin Receptor 1B, Mouse, Unlabeled, Clone: 1C2.1,

Catalog No. MABN1539MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1539MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1539MI Supplier MilliporeSigma™ Supplier No. MABN1539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-hydroxytryptamine receptor 1B (UniProt P28222; also known as 5-HT-1B, 5-HT-1D-beta, 5-HT1B, S12, Serotonin 1D beta receptor, Serotonin receptor 1B) is encoded by the HTR1B (also known as HTR1DB) gene (Gene ID 3351) in human. The 5-HT receptors are grouped according to their antagonist susceptibilities and their affinities for 5-HT. The 5-HT1 receptors are a subfamily of 5-HT receptors that were originally classified according to their inhibition of adenylyl cyclase, degree of sequence similarity and their overlapping pharmacological specificities. The subfamily has five different receptors: 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E, 5-HT1F that can couple to Gi/Go and mediate inhibitory neurotransmission. 5HT-1B receptor belong to the family of G-protein coupled receptors that are distributed in brain cortex, striatum, amygdala, medulla, hippocampus, caudate nucleus and putamen. 5-HT-1B also functions as a receptor for ergot alkaloid derivatives, various anxiolytic and antidepressant drugs and other psychoactive substances, such as lysergic acid diethylamide. Ligand binding causes a conformation change in the receptor that triggers signaling via G-proteins, which modulates the activity of down-stream effectors, such as adenylate cyclase. Signaling via 5-HT-1B inhibits adenylate cyclase activity. 5-HT1B receptor is known to be involved in thermoregulation, respiration, appetite control, sexual behavior, and aggression. 5-HT-1B receptor are therapeutic targets for antimigraine drugs and for treatment of bipolar disorders, autism, and anti-aggressive agents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin Receptor 1B
Applications Western Blot
Classification Monoclonal
Clone 1C2.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene HTR1B, HTR1DB
Gene Accession No. P28222
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to the thrid cytoplasmic domain of human serotonin receptor 1B.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000854
Test Specificity Clone 1C2.1 targets an epitope within the third cytoplasmic domain of human, mouse, and rat serotonin receptor 1B.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.