Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Serotonin receptor 3A/HTR3A, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1475MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1475MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1475MI Supplier MilliporeSigma™ Supplier No. ABN1475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-hydroxytryptamine receptor 3A (UniProt P46098; also known as 5-HT-3, 5-HT3-A, 5-HT3A, 5-HT3R, 5-hydroxytryptamine receptor 3, Serotonin receptor 3A, Serotonin-gated ion channel receptor) is encoded by the HTR3A (also known as 5HT3R, HTR3) gene (Gene ID 3359) in human. Hydroxytryptamine (5-HT) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT. 5-hydroxytryptamine receptor 3 (5-HT3R) is a ligand-gated ion channel involved in many neurophysiological functions and is widely expressed throughout the brain and in peripheral organs. Five different subunits (A through E) have been identified, with the homopentameric 5-HT3A being the most studied form. 5-HT3A is produced with a signal peptide sequence (a.a. 1-23), which is removed posttranslationally to yield the mature 4-transmembrane (a.a. 242-268, 274-292, 303-321, 456-475) ion channel with both its N- and C-terminal ends exposed extracellularly (a.a. 24-241 and 476-478).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serotonin receptor 3A/HTR3A
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene HTR3A, 5HT3R, HTR3
Gene Accession No. P46098
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence from the second cytomplasmic domain of human Serotonin receptor 3A/HTR3A.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000860
Test Specificity This rabbit polyclonal antibody detects human Serotonin receptor 3A/HTR3A by targeting a sequence within the second cytoplasmic domain.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.