Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550255
Description
The antibody detects endogenous levels of total stress-associated endoplasmic reticulum protein 1 protein.
This gene interacts with target proteins during their translocation into the lumen of the endoplasmic reticulum. Protects unfolded target proteins against degradation during ER stress. May facilitate glycosylation of target proteins after termination of ER stress. May modulate the use of N-glycosylation sites on target proteins.
Specifications
SERP1 | |
Polyclonal | |
Unconjugated | |
SERP1 | |
D3Ucla1; RAMP4; ribosome associated membrane protein 4; ribosome-attached membrane protein 4; Serp1; Stress-associated endoplasmic reticulum protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27230, 28146, 80881 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9R2C1, Q9Y6X1, Q9Z1W5 | |
SERP1 | |
Fusion protein corresponding to residues near the N terminal of human stress-associated endoplasmic reticulum protein 1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction