Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558615 Supplier Invitrogen™ Supplier No. PA558615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAKQRIRMAN EKHSKNITQR GNVAKT Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene interacts with target proteins during their translocation into the lumen of the endoplasmic reticulum. Protects unfolded target proteins against degradation during ER stress. May facilitate glycosylation of target proteins after termination of ER stress. May modulate the use of N-glycosylation sites on target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SERP1
Gene Accession No. Q9R2C1, Q9Y6X1, Q9Z1W5
Gene Alias D3Ucla1; RAMP4; ribosome associated membrane protein 4; ribosome-attached membrane protein 4; Serp1; Stress-associated endoplasmic reticulum protein 1
Gene Symbols SERP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERP1. Recombinant protein control fragment (Product #RP-104317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27230, 28146, 80881
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.