Learn More
Eisco™ Serpentinite Metamorphic Rock Specimen
SCI_ED
Catalog No. S27614 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27614 12/Pk.
S27592 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27614 Supplier Eisco™ Supplier No. ESNG0085PK12
Add to Cart
Add to Cart
When clay- and aluminum-rich rock is altered by water from geothermal systems, serpentinite is liable to form. Serpentinite is often found in low-grade metamorphic areas and is formed when clay and aluminum rich rock is altered by water from geothermal systems.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Greenish in color and smooth to the touch
  • Often found as a plane of weakness in faulted rocks
  • Excellent for lessons on metamorphic rocks, low-grade deformation, and structural geology
  • Serpentinite is composed of hydrated silicate minerals, particularly phyllosilicates
Specifications
Product Type Metamorphic Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 2.4 kg