Learn More
Eisco™ Serpentinite Metamorphic Rock Specimen
SCI_ED
Catalog No. p-7203017 Shop All Eisco Products
Click to view available options
Quantity:
1/Ea.
12/Pk.
When clay- and aluminum-rich rock is altered by water from geothermal systems, serpentinite is liable to form. Serpentinite is often found in low-grade metamorphic areas and is formed when clay and aluminum rich rock is altered by water from geothermal systems.
  • Correlates well to Next Generation Science Standards Evidence
  • Ideal for teaching students about rounding and sorting, as well as for identification & classification exercises
  • Greenish in color and smooth to the touch
  • Often found as a plane of weakness in faulted rocks
  • Excellent for lessons on metamorphic rocks, low-grade deformation, and structural geology
  • Serpentinite is composed of hydrated silicate minerals, particularly phyllosilicates
Specifications
Product Type Metamorphic Rock
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 2.4 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.