Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Antibody (005), Alexa Fluor™ 532

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290930AF532
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Rat samples. It is validated for ELISA.Specifications
Serpin A1/alpha 1-Antitrypsin | |
Monoclonal | |
Alexa Fluor 532 | |
A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Serpin A1/alpha 1-Antitrypsin (NP_071964.2; Met1-Arg411). | |
0.1 mL | |
Stem Cell Markers | |
5265 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
5 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction