Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Antibody (OTI9A1), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274095AF647
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Serpin A1/alpha 1-Antitrypsin | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SERPINA1 | |
Full-length protein expressed in 293T cell transfected with human Serpin A1/alpha 1-Antitrypsin expression vector. | |
0.1 mL | |
Primary | |
Human, Canine, Primate, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI9A1 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
5265.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction