Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A11 Antibody (202), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290584IR
Description
Serpin A11 Monoclonal specifically detects Serpin A11 in Mouse samples. It is validated for ELISA.Specifications
Serpin A11 | |
Monoclonal | |
DyLight 755 | |
antiproteinase-like 2, antitrypsin), member 11, member 11, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A11, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin A11 (NP_955018.2; Met 1-Gly 424). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
202 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
256394 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction