Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A3/alpha 1-Antichymotrypsin Antibody (006), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289404H
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Serpin A3/alpha 1-Antichymotrypsin | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human SerpinA7 Human SerpinA5 Human SerpinC1 Human SerpinD1 Human SerpinF2 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
6 | |
ELISA, Sandwich ELISA Detection | |
AACTMGC88254, ACTGIG25, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A3/alpha 1-Antichymotrypsin (NP_001076.2; Met 1-Ala 423). | |
0.1 mL | |
Breast Cancer, Cancer | |
12 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction