Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A5/Protein C Inhibitor Antibody (110), DyLight 680 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289406FR
Description
Serpin A5/Protein C Inhibitor Monoclonal antibody specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for ELISASpecifications
Serpin A5/Protein C Inhibitor | |
Monoclonal | |
DyLight 680 | |
Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A5/Protein C Inhibitor (NP_000615.3; Met 1-Pro 406). | |
0.1 mL | |
Cancer | |
5104 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
110 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction