Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A5/Protein C Inhibitor Antibody (OTI1C4), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274100APCCY7
Description
Serpin A5/Protein C Inhibitor Monoclonal specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Flow Cytometry.Specifications
Serpin A5/Protein C Inhibitor | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
SERPINA5 | |
Full length recombinant protein of human Serpin A5/Protein C Inhibitor (NP_000615) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI1C4 | |
Flow Cytometry | |
Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
Mouse | |
Affinity Purified | |
Cancer | |
5104.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction