Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (014), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290529AF405
Description
Serpin A6/Cortisol Binding Globulin Monoclonal specifically detects Serpin A6/Cortisol Binding Globulin in Mouse samples. It is validated for ELISA.Specifications
Serpin A6/Cortisol Binding Globulin | |
Monoclonal | |
Alexa Fluor 405 | |
antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin A6/Cortisol Binding Globulin (Q06770; Met1-Ala397). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human SerpinA1 Human SerpinA3 Human SerpinA5 Human Serpinb6 Mouse SerpinB8 Human Serpinb9 Mouse SerpinD1 Mouse SerpinF1 Human SerpinG1 Mouse SerpinI1 Human SerpinI2 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
14 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
866 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction