Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (06), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306555UV
Description
Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for ELISASpecifications
Serpin A6/Cortisol Binding Globulin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
866 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
06 | |
ELISA | |
antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Serpin A6/Cortisol Binding Globulin (NP_001747.2; Met1-Val405). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction