Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Antibody (SAIC-42B-10), Alexa Fluor™ 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320108AF405
Description
Serpin A6/Cortisol Binding Globulin Monoclonal antibody specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for Western BlotSpecifications
Serpin A6/Cortisol Binding Globulin | |
Monoclonal | |
Alexa Fluor 405 | |
antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
Peptide "e;AQLLQGLGFNLTER"e; derived from the Corticosteroid-binding globulin SERPINA6, conjugated to KLH. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
SAIC-42B-10 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
866 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction