Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A6/Cortisol Binding Globulin Rabbit anti-Human, Clone: 37, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289739100UL
Description
Serpin A6/Cortisol Binding Globulin Monoclonal specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for ELISA.Specifications
Serpin A6/Cortisol Binding Globulin | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
866 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
37 | |
ELISA 1:5000-1:10000 | |
antitrypsin), member 6, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
Recombinant Human Serpin A6/Cortisol Binding Globulin protein | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction