Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A7/TBG Antibody (010), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289735AF405
Description
Serpin A7/TBG Monoclonal antibody specifically detects Serpin A7/TBG in Human samples. It is validated for ELISASpecifications
Serpin A7/TBG | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
6906 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
010 | |
ELISA | |
antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A7/TBG (NP_000345.2; Met 1-Ala 415). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction