Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A8/Angiotensinogen Antibody (001), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290541B
Description
Serpin A8/Angiotensinogen Monoclonal specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot.Specifications
Serpin A8/Angiotensinogen | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
1 | |
Western Blot | |
alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin A8/Angiotensinogen (P11859; Met1-Val477). | |
0.1 mL | |
Lipid and Metabolism | |
183 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction