Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin A8/Angiotensinogen Rabbit anti-Mouse, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Serpin A8/Angiotensinogen |
---|---|
Clone | 1 |
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Serpin A8/Angiotensinogen Monoclonal specifically detects Serpin A8/Angiotensinogen in Mouse samples. It is validated for Western Blot.Specifications
Serpin A8/Angiotensinogen | |
Western Blot 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
alpha-1 antiproteinase, antitrypsin, angiotensin II, angiotensinogen, angiotensinogen (serpin peptidase inhibitor, clade A, member 8), ANHU, FLJ92595, FLJ97926, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8, SERPINA8angiotensin I | |
Recombinant Mouse Serpin A8/Angiotensinogen Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
183 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title