Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin B13 Mouse anti-Human, Mouse, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABS132025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1320-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1320-25 Supplier MilliporeSigma Supplier No. MABS132025UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Serpin B13, clone B29, Cat. No. MABS1320, is a mouse monoclonal antibody that detects Serpin B13 and has been tested for use in Function Studies, Immunofluorescence, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serpin B13
Applications Functional Assay, Immunofluorescence, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Symbols Serpinb13
Host Species Mouse
Immunogen Purified full length mouse Serpin B13 resuspended in Complete Freund's Adjuvant.
Molecular Weight of Antigen 44.32 kda calculated.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone B29 detects Serpin B13 in human and murine species.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.