Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B3/SCCA1 Antibody (CPTC-SERPINB3-2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308536R
Description
Serpin B3/SCCA1 Monoclonal antibody specifically detects Serpin B3/SCCA1 in Human samples. It is validated for Western Blot, Protein ArraySpecifications
Serpin B3/SCCA1 | |
Monoclonal | |
DyLight 550 | |
HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
Recombinant human full-length Serpin B3/SCCA1 protein (Uniprot: P29508) | |
100 μg | |
Cancer | |
6317 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Protein Array | |
CPTC-SERPINB3-2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction