Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B3/SCCA1 Antibody (OTI3C2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274108C
Description
Serpin B3/SCCA1 Monoclonal specifically detects Serpin B3/SCCA1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Serpin B3/SCCA1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SERPINB3 | |
Full length recombinant protein of human Serpin B3/SCCA1 (NP_008850) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3C2 | |
Western Blot, Immunohistochemistry | |
HsT1196, Protein T4-A, SCC, SCCA, SCCA1SCCA-1, SCCA-PD, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3, serpin B3, serpin peptidase inhibitor, clade B (ovalbumin), member 3, Squamous cell carcinoma antigen 1, T4-A | |
Mouse | |
Affinity Purified | |
Cancer | |
6317.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction