Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B5/Maspin Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11097377G
Description
Serpin B5/Maspin Polyclonal specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Knockdown Validated.Specifications
Serpin B5/Maspin | |
Polyclonal | |
Western Blot 0.5 ug/ml, Knockdown Validated | |
P36952 | |
SERPINB5 | |
Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot# P36952] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, KnockDown | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
Rabbit | |
40 kDa | |
0.1 mL | |
Cancer, Tumor Suppressors | |
5268 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction