Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B5/Maspin Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11097377MFV450
Description
Serpin B5/Maspin Polyclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, ICC/IF (Negative), KnockDownSpecifications
Serpin B5/Maspin | |
Polyclonal | |
Western Blot, ICC/IF (Negative), Knockdown Validated | |
maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot# P36952] | |
0.1 mL | |
Biologically Active Proteins, Cancer, Tumor Suppressors | |
5268 | |
Store at 4C in the dark. | |
IgG |
Western Blot, KnockDown | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction