Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B5/Maspin Mouse anti-Human, Clone: SERPINB5/4972, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Serpin B5/Maspin |
---|---|
Clone | SERPINB5/4972 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
Serpin B5/Maspin Monoclonal antibody specifically detects Serpin B5/Maspin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Serpin B5/Maspin | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Cancer, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
maspin, Peptidase inhibitor 5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5 | |
Recombinant fragment of human Serpin B5/Maspin protein (around aa1-200) (exact sequence is proprietary) (Uniprot: P36952) | |
Primary | |
Store at 4°C. |
SERPINB5/4972 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P36952 | |
5268 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title