Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin B8/Proteinase Inhibitor 8 Antibody (013), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290484AF750
Description
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Mouse samples. It is validated for ELISA.Specifications
Serpin B8/Proteinase Inhibitor 8 | |
Monoclonal | |
Alexa Fluor 750 | |
CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin B8/Proteinase Inhibitor 8 (NP_035589.1; Met 1-Pro 374). | |
0.1 mL | |
Cardiovascular Biology | |
5271 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
13 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction