Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin C1/Antithrombin-III Antibody (023), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289296PCP
Description
Serpin C1/Antithrombin-III Monoclonal antibody specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for ELISASpecifications
| Serpin C1/Antithrombin-III | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 023 | |
| ELISA | |
| AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin C1/Antithrombin-III (Accession#: NP_000479.1; Met 1-Lys 464). | |
| 0.1 mL | |
| Cardiovascular Biology, Stem Cells | |
| 462 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction