Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin C1/Antithrombin-III Antibody (OTI8D5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274113
Description
Serpin C1/Antithrombin-III Monoclonal specifically detects Serpin C1/Antithrombin-III in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Serpin C1/Antithrombin-III | |
Monoclonal | |
LYOPH | |
Flow Cytometry 1:100, CyTOF-ready | |
AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
Mouse | |
52.6 kDa | |
100 μg | |
462 | |
Human, Mouse, Rat | |
Lyophilized |
Flow Cytometry, CyTOF | |
OTI8D5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SERPINC1 | |
Full length human recombinant protein of human SERPINC1 (NP_000479) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction