Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin C1/Antithrombin-III Antibody (SR1949), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Serpin C1/Antithrombin-III |
---|---|
Clone | SR1949 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
Serpin C1/Antithrombin-III Monoclonal antibody specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Serpin C1/Antithrombin-III | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AT3antithrombin-III, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1 | |
A synthesized peptide derived from human Serpin C1/Antithrombin-III (Uniprot #: P01008) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1949 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Stem Cells | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
462 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title