Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin E1/PAI-1 Antibody (004), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289393AF405
Description
Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Serpin E1/PAI-1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
4 | |
ELISA, Sandwich ELISA Detection | |
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin E1/PAI-1 (NP_000593.1; Met 1-Pro 402). | |
0.1 mL | |
Cancer | |
5054 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction