Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin E1/PAI-1 Antibody (1C5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259495H
This item is not returnable.
View return policy
Description
Serpin E1/PAI-1 Monoclonal antibody specifically detects Serpin E1/PAI-1 in Human samples. It is validated for Western Blot, ELISASpecifications
Serpin E1/PAI-1 | |
Monoclonal | |
HRP | |
Endothelial plasminogen activator inhibitor, PAI1PAI-1, PAISerpin E1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
Recombinant human Serpin E1/PAI-1 (24-402aa) purified from E. coli. | |
0.1 mL | |
Cancer | |
5054 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1C5 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction