Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin E2/PN1 Antibody (OTI2C9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274117F
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
Serpin E2/PN1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SERPINE2 | |
Full length recombinant protein of human Serpin E2/PN1 (NP_006207) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI2C9 | |
Flow Cytometry, Immunohistochemistry | |
GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
5270.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction