Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F1/PEDF Antibody (013), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289798F
Description
Serpin F1/PEDF Monoclonal antibody specifically detects Serpin F1/PEDF in Human samples. It is validated for ELISASpecifications
Serpin F1/PEDF | |
Monoclonal | |
FITC | |
Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin F1/PEDF (Met 1-Pro 418; NP_002606.3) . | |
0.1 mL | |
Biologically Active Proteins, Neuroscience, Vision | |
5176 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
013 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction