Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F1/PEDF Antibody (OTI1F7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201868
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human, Mouse samples. It is validated for Western Blot.Specifications
| Serpin F1/PEDF | |
| Monoclonal | |
| 1.08 mg/mL | |
| Western Blot 1:1000 | |
| SERPINF1 | |
| Full length human recombinant protein of human SERPINF1(NP_002606) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a |
| Western Blot | |
| OTI1F7 | |
| Unconjugated | |
| Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| Mouse | |
| 46.1 kDa | |
| 0.1 mL | |
| Neuroscience, Vision | |
| 5176 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction