Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F1/PEDF Antibody (SR1798), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Serpin F1/PEDF |
---|---|
Clone | SR1798 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
Serpin F1/PEDF Monoclonal antibody specifically detects Serpin F1/PEDF in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
Serpin F1/PEDF | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
A synthesized peptide derived from human Serpin F1/PEDF (Uniprot #: P36955) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1798 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Neuroscience, Vision | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5176 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title