Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F1/PEDF Rabbit anti-Human, Mouse, Rat, Clone: 4H0C5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Serpin F1/PEDF |
---|---|
Clone | 4H0C5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Serpin F1/PEDF Monoclonal antibody specifically detects Serpin F1/PEDF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Serpin F1/PEDF | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Neuroscience, Vision | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5176 | |
IgG | |
Affinity purified |
4H0C5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Serpin F1/PEDF (P36955). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title