Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin F2/alpha 2-Antiplasmin Antibody (RM0324-2H63), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211890AF532
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blot.Specifications
Serpin F2/alpha 2-Antiplasmin | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SERPINF2 | |
Mouse recombinant Serpin F2/alpha 2-Antiplasmin | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot | |
RM0324-2H63 | |
Western Blot | |
AAPclade F (alpha-2 antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
Rat | |
Protein A or G purified | |
Cardiovascular Biology | |
5345 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction