Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin G1/C1 Inhibitor Antibody (18), DyLight 550, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP289738R

Catalog No. NB109038


Only null left
Add to Cart

Description

Description

Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.
Specifications

Specifications

Serpin G1/C1 Inhibitor
Monoclonal
DyLight 550
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin G1/C1 Inhibitor (NP_000053.2; Met 1-Ala 500).
0.1 mL
Primary
No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ SERPINA3/ AACT Human SerpinA5/ PROCI Human SerpinC1/SERPINC1/ Antithrombin-III Human SerpinD1/ SERPIND1/ HC2 Human SerpinF1/ SERPINF1/ PEDF Human SerpinF2/ SERPINF2 Human SerpinI1/ SERPINI1/ Protease inhibitor 12
Store at 4°C in the dark.
IgG
Western Blot, ELISA
18
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
710
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.