Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin G1/C1 Inhibitor Antibody (18), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289738JF549
Description
Serpin G1/C1 Inhibitor Monoclonal specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Serpin G1/C1 Inhibitor | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
710 | |
Human | |
Purified |
ELISA | |
18 | |
ELISA, Sandwich ELISA Detection | |
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin G1/C1 Inhibitor (NP_000053.2; Met 1-Ala 500). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human SerpinA1/ A1AT Human SerpinA3/ SERPINA3/ AACT Human SerpinA5/ PROCI Human SerpinC1/SERPINC1/ Antithrombin-III Human SerpinD1/ SERPIND1/ HC2 Human SerpinF1/ SERPINF1/ PEDF Human SerpinF2/ SERPINF2 Human SerpinI1/ SERPINI1/ Protease inhibitor 12 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction