Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin G1/C1 Inhibitor Antibody, Alexa Fluor™ 405, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP311027AF405
Description
Serpin G1/C1 Inhibitor Polyclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human samples. It is validated for Western Blot, ELISASpecifications
Serpin G1/C1 Inhibitor | |
Polyclonal | |
Western Blot, ELISA | |
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
Human Serpin G1/C1 Inhibitor | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Sheep | |
Affinity purified | |
RUO | |
710 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction