Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin G1/C1 Inhibitor Antibody (HL1196) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325387
Description
Serpin G1/C1 Inhibitor Monoclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Serpin G1/C1 Inhibitor | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
HL1196 | |
Western Blot 1:500-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
Recombinant protein encompassing a sequence within the center region of human Serpin G1/C1 Inhibitor. The exact sequence is proprietary. | |
100 μL | |
Cardiovascular Biology, Immunology, Innate Immunity | |
710 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction