Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin G1/C1 Inhibitor Antibody (SR1022), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322363100UL
Description
Serpin G1/C1 Inhibitor Monoclonal antibody specifically detects Serpin G1/C1 Inhibitor in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Serpin G1/C1 Inhibitor | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1022 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
C1IN, C1INH, C1NH, HAE1, HAE2, SERPING1 serpin peptidase inhibitor, clade G (C1 inhibitor), member 1 | |
A synthesized peptide derived from human Serpin G1/C1 Inhibitor (Uniprot #: P05155) | |
100 μg | |
Cardiovascular Biology, Immunology, Innate Immunity | |
710 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction