Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serpin I1/Neuroserpin Antibody (002), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290745MFV450
Description
Serpin I1/Neuroserpin Monoclonal antibody specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Serpin I1/Neuroserpin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
002 | |
Immunohistochemistry-Paraffin | |
DKFZp781N13156, neuroserpin, Peptidase inhibitor 12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin I1/Neuroserpin (Uniprot#: O35684; Met1-Leu410). | |
0.1 mL | |
Biologically Active Proteins | |
5274 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction