Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121068 Supplier Invitrogen™ Supplier No. PA5121068
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, Mouse liver, Rat liver. The target is usually found in the following locations: Secreted. Immunogen sequence: FAFIHKDFDV KETFFNLSKR YFDTECVPMN FRNASQAKRL MNHYINKETR GKIPKLFDEI NPETKLILVD YILFKGKWLT PFDPVFTEVD TFHLDKYKTI KVPMMYGAGK FASTFDKNFR CHVLKLPYQG NATMLVVLME KMGDHLALED YLTTDLVETW LRNMKTRNME VFFPKFKLDQ KYEMHELLRQ MGIRRIFSPF ADLSELSATG RNLQVSRVLQ RTVIEVDERG TEAVAGILSE ITAYSMPPVI KVDRPFHFMI YEETSGMLLF LGRVVNPTLL.

The protein encoded by this gene belongs to the serpin family. It is predominantly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SERPINA10
Gene Accession No. Q62975, Q8R121, Q9UK55
Gene Alias plasma protein associated with liver regeneration; protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; PZI; Rasp1; RASP-1; Regeneration-associated serpin 1; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin A10; serpin family A member 10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin peptidase inhibitor, clade A, member 10; Serpina10; UNQ707/PRO1358; Zpi
Gene Symbols SERPINA10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-444 of human SERPINA10 (NP_001094077.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171154, 217847, 51156
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.