Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SERPINA10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583924 Supplier Invitrogen™ Supplier No. PA583924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CHVLKLPYQG NATMLVVLME KMGDHLALED YLTTDLVETW LRNMKTRNME VFFPKFKLDQ KYEMHELLRQ MGIRRIFSPF ADLSELSATG RNLQVSRVLQ RTVIEVDERG TEAVAGILSE ITAYSMPPVI KVDRPFHFMI YEETSGMLLF L Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.

The protein encoded by this gene belongs to the serpin family. It is predominantly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERPINA10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SERPINA10
Gene Accession No. Q9UK55
Gene Alias plasma protein associated with liver regeneration; protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; PZI; Rasp1; RASP-1; Regeneration-associated serpin 1; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin A10; serpin family A member 10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin peptidase inhibitor, clade A, member 10; Serpina10; UNQ707/PRO1358; Zpi
Gene Symbols SERPINA10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SERPINA10. Recombinant protein control fragment (Product #RP-88933).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.