Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERPINA11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598585
Description
SERPINA11 Polyclonal Antibody for Western Blot, ELISA
The protein belongs to the serpin family. SERPINA11 has serine-type endopeptidase inhibitor activity, negative regulation of endopeptidase activity, regulation of proteolysis, serine-type endopeptidase inhibitor activity.
Specifications
SERPINA11 | |
Polyclonal | |
Unconjugated | |
SERPINA11 | |
antiproteinase-like 2; Gm895; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; serine (or cysteine) proteinase inhibitor, clade A, member 11; serpin A11; serpin family A member 11; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11; Serpina11 | |
Rabbit | |
Protein G | |
RUO | |
256394 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.16 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86U17 | |
SERPINA11 | |
Recombinant Human Serpin A11 protein (279-394AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction